missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAB1 Antibody (OTI4G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00480
This item is not returnable.
View return policy
Description
NECAB1 Monoclonal specifically detects NECAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| NECAB1 | |
| Monoclonal | |
| 0.59 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100 | |
| NECAB1 | |
| Full length human recombinant protein of human NECAB1 (NP_071746) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 64168 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry | |
| OTI4G8 | |
| Unconjugated | |
| EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1 | |
| Mouse | |
| 40.4 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction