missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35564-100ul
This item is not returnable.
View return policy
Description
NECAB3 Polyclonal antibody specifically detects NECAB3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| NECAB3 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| Amyloid beta A4 protein-binding family A member 2-binding protein, APBA2BPfamily A, member 2 binding protein, dJ63M2.4, dJ63M2.5, EFCBP3STIP3, EF-hand calcium binding protein 3, Neuronal calcium-binding protein 3, neuronal calcium-binding protein NECAB3, NIP1Nek2-interacting protein 1, N-terminal EF-hand calcium binding protein 3, N-terminal EF-hand calcium-binding protein 3, synaptotagmin interacting protein 2, synaptotagmin interacting protein STIP3, SYTIP2, XB51X11L-binding protein 51 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-362 of human NECAB3 (NP_112508.3).,, Sequence:, MACAGLLTVCLLRPPAPQPQPQTPRHPQLAPDPGPAGHTLFQDVFRRADKNDDGKLSFEEFQNYFADGVLSLGELQELFSGIDGHLTDNLETEKLCDYFSEHLGVYRPVLAALESLNRAVLAAMDATKLEYERASKVDQFVTRFLLRETVSQLQALQSSLEGASDTLEAQAHGWRSDAESVEAQSRLCGSRRAGRRALRSVSRSSTWSPGSSDTGRSSEAEMQWRLQVNRLQELIDQLECKAPRLEPLREEDLAKGPDLHILMAQRQVQVAEEGLQDFHRALRCYVDFTGAQSHCLHVSAQKMLDGASFTLYEFWQDEASWRRHQQSPGSKAFQRILIDHLRAPDTLTTVFFPASWWIMNNN | |
| 100 μL | |
| Lipid and Metabolism | |
| 63941 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction