missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAP2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97285B
This item is not returnable.
View return policy
Description
NECAP2 Polyclonal antibody specifically detects NECAP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NECAP2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Biotin | |
| adaptin ear-binding coat-associated protein 2, adaptin-ear-binding coat-associated protein 2, FLJ10420, NECAP endocytosis associated 2, NECAP endocytosis-associated protein 2, NECAP-2 | |
| Produced in rabbits immunized with E. coli-derived Human NECAP2 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 55707 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction