missing translation for 'onlineSavingsMsg'
Läs mer

NECAP2 Antibody, Novus Biologicals™

Produktkod. p-200065342 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
100 μg
50 μg
Förpackningsstorlek:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
18625542 100 μg 100µL
18633565 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18625542 Leverantör Novus Biologicals Leverantörsnummer NBP297285100ul

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

NECAP2 Polyclonal antibody specifically detects NECAP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen NECAP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.0)
Gene Alias adaptin ear-binding coat-associated protein 2, adaptin-ear-binding coat-associated protein 2, FLJ10420, NECAP endocytosis associated 2, NECAP endocytosis-associated protein 2, NECAP-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NECAP2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55707
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.