missing translation for 'onlineSavingsMsg'
Learn More

Necdin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-57334-25ul

 View more versions of this product

Product Code. 18683517

  • ONLINE EXCLUSIVE PRICE
    € 415.00
    € 391.65 / 25µL
    Save € 23.35
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Necdin Polyclonal specifically detects Necdin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Necdin
Polyclonal
Immunocytochemistry/Immunofluorescence 1-4 μg/mL
HsT16328, necdin, necdin (mouse) homolog, necdin homolog (mouse), PWCR
Rabbit
Affinity Purified
RUO
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Immunocytochemistry, Immunofluorescence
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
NDN
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AQLVQKAHELMWYVLVKDQKKMIIWFPDMVKDVIGSYKKWCRSILRRTSLI
25 μL
Apoptosis, Cancer, Tumor Suppressors
4692
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only