missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Necdin Antibody (OTI5D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46117
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
Necdin Monoclonal specifically detects Necdin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifications
| Necdin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
| HsT16328, necdin, necdin (mouse) homolog, necdin homolog (mouse), PWCR | |
| Mouse | |
| 35.9 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 4692 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI5D1 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| NDN | |
| Full length human recombinant protein of human NDN(NP_002478) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction