missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Necdin Antibody (OTI5D1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72925PCP
This item is not returnable.
View return policy
Description
Necdin Monoclonal antibody specifically detects Necdin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Necdin | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI5D1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| HsT16328, necdin, necdin (mouse) homolog, necdin homolog (mouse), PWCR | |
| Full length human recombinant protein of human NDN(NP_002478) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 4692 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction