missing translation for 'onlineSavingsMsg'
Learn More

Necrostatin-1, TRC

Product Code. 30476524
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30476524 10 mg
30483890 50 mg
30480354 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476524 Supplier Toronto Research Chemicals Supplier No. TRCN38855010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4311-88-0
Molecular Formula C13 H13 N3 O S
Molecular Weight (g/mol) 259.33
Synonym 5-(1H-Indol-3-ylmethyl)-3-methyl-2-thioxo-4-imidazolidinone
IUPAC Name 5-(1H-indol-3-ylmethyl)-3-methyl-2-sulfanylideneimidazolidin-4-one
SMILES CN1C(=S)NC(Cc2c[nH]c3ccccc23)C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H13N3OS/c1-16-12(17)11(15-13(16)18)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,11,14H,6H2,1H3,(H,15,18)
Formula Weight 259.0779
Chemical Name or Material Necrostatin-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.