missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-2/CD112 Antibody (829038), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3869B
This item is not returnable.
View return policy
Description
Nectin-2/CD112 Monoclonal antibody specifically detects Nectin-2/CD112 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Nectin-2/CD112 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 829038 | |
| Flow Cytometry, CyTOF-ready | |
| CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nectin-2/CD112, Gln32-Gly351 (predicted), Accession # P32507 | |
| 0.1 mL | |
| Immunology | |
| 5819 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction