missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hHR23b Antibody (SAIC-28A-29), HRP , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20096H
This item is not returnable.
View return policy
Description
hHR23b Monoclonal antibody specifically detects hHR23b in Human samples. It is validated for Western Blot
Specifications
| hHR23b | |
| Monoclonal | |
| HRP | |
| HHR23B, HR23BUV excision repair protein RAD23 homolog B, p58, RAD23 (S. cerevisiae) homolog B, RAD23 homolog B (S. cerevisiae), RAD23, yeast homolog of, B, XP-C repair complementing complex 58 kDa, XP-C repair complementing protein, XP-C repair-complementing complex 58 kDa protein | |
| Peptide "e;ILNDDTALK"e;, derived from the UV excision repair protein RAD23 homolog B, conjugated to KLH. | |
| 0.1 mL | |
| Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers, Tumor Suppressors | |
| 5887 | |
| Store at 4&drg;C in the dark. | |
| IgG κ |
| Western Blot | |
| SAIC-28A-29 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction