missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-4/PVRL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20014
This item is not returnable.
View return policy
Description
Nectin-4/PVRL4 Polyclonal specifically detects Nectin-4/PVRL4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Nectin-4/PVRL4 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (92%), Rat (94%), Bovine (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.02mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q96NY8 | |
| NECTIN4 | |
| Recombinant protein encompassing a sequence within the center region of human PVRL4. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 81607 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction