missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nectin 4 Recombinant Human Monoclonal Antibody (6D3)

Product Code. 17958822
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17958822 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17958822 Supplier Invitrogen™ Supplier No. MA549975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Nectin 4 Recombinant Monoclonal Antibody for ELISA

Nectin 4 (PVRL4) is a gene located on chromosome 1q23.3 that encodes a member of the nectin family of cellular adhesion molecules. These proteins are primarily involved in forming adherens junctions between epithelial cells, contributing to tissue integrity and maintenance in various tissues, including the skin, lungs, and mammary glands. Nectin 4 is highly expressed during embryonic development and is crucial for processes like tissue morphogenesis and repair. It acts as a ligand for the poliovirus receptor-related family and participates in cell signaling pathways that influence cell proliferation and survival. In the context of cancer, Nectin 4 is often upregulated, particularly in breast, ovarian, and lung cancers, where its expression is associated with tumor progression, metastasis, and poor prognosis. This makes Nectin 4 a potential target for therapeutic interventions and a candidate biomarker for cancer diagnosis and monitoring.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nectin 4
Applications ELISA
Classification Recombinant Monoclonal
Clone 6D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene NECTIN4
Gene Accession No. Q96NY8
Gene Alias 1200017F15Rik; EDSS1; ig superfamily receptor LNIR; LNIR; nectin 4; nectin cell adhesion molecule 4; NECTIN4; Nectin-4; poliovirus receptor-related 4; poliovirus receptor-related protein 4; Processed poliovirus receptor-related protein 4; PRR4; Pvrl4; RGD1559826
Gene Symbols NECTIN4
Host Species Human
Immunogen Recombinant Human NECTIN4 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81607
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.