missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEDD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558455
This item is not returnable.
View return policy
Description
Immunogen sequence: SSTPNPKIAS SVTAGVASSL SEKIADSIGN NRQNAPLTSI QIRFIQNMIQ ETLDDFREAC HRDIVNLQVE MIKQFHMQLN EMHSLLERYS VNEGL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.
Specifications
| NEDD1 | |
| Polyclonal | |
| Unconjugated | |
| Nedd1 | |
| GCP-WD; Nedd1; Nedd-1; NEDD1 gamma-tubulin ring complex targeting factor; neural precursor cell expressed developmentally down-regulated protein 1; neural precursor cell expressed, developmentally down-regulated 1; neural precursor cell expressed, developmentally down-regulated gene 1; protein NEDD1; TUBGCP7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121441 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NHV4 | |
| Nedd1 | |
| Recombinant protein corresponding to Human NEDD1. Recombinant protein control fragment (Product #RP-97798). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction