missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEDD4 Monoclonal Antibody (K24022_16E6), Invitrogen™
Mouse Monoclonal Antibody
€ 450.00 - € 676.00
Specifications
| Antigen | NEDD4 |
|---|---|
| Clone | K24022_16E6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30197611
|
Invitrogen™
MA555813 |
50 μL |
€ 450.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30197698
|
Invitrogen™
MA556193 |
200 μL |
€ 676.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: NARLTIFGNS AVSQPASSSN HSSRRGSLQA YTFEEQPTLP VLLPTSSGLP PGWEEKQDER GRSYYVDHNS RTTTWTKPTV QATVETSQLT SSQSSAGPQS QASTSDSGQQ
NEDD4 Monoclonal antibody specifically detects NEDD4 in Human samples. It is validated for Western BlotSpecifications
| NEDD4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AA959633; AL023035; AU019897; cell proliferation-inducing gene 53 protein; E3 ubiquitin-protein ligase NEDD4; E430025J12Rik; HECT-type E3 ubiquitin transferase NEDD4; Kiaa0093; mKIAA0093; NEDD4; NEDD-4; NEDD4 E3 ubiquitin protein ligase; Nedd4-1; Nedd4a; Neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated 4; neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4a; neural precursor cell expressed, developmentally down-regulted gene 4; OTTHUMP00000217840; OTTHUMP00000217841; OTTHUMP00000217842; PIG53; receptor-potentiating factor 1; RPF1 | |
| Recombinant human NEDD4 (immunogen range is 721-830). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| NEDD4 |
| K24022_16E6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P46934 | |
| 4734 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title