missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZB Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-814
This item is not returnable.
View return policy
Description
CAPZB Polyclonal specifically detects CAPZB in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| CAPZB | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Cap Z, CAPB, CAPPB, capping protein (actin filament) muscle Z-line, beta, CAPZ, CapZ beta, F-actin capping protein beta subunit, F-actin-capping protein subunit beta, MGC104401, MGC129749, MGC129750 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 832 | |
| Human, Mouse, Rat, Pig | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_004921.1 | |
| CAPZB | |
| Peptide with sequence NDLVEALKRKQQC corresponding to C-Terminus according to NP_004921.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to be specific for the beta2 isoform of capping protein, found in non-muscle cells and should not recognize the beta1 form. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction