missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total NEDD8.
NEDD8 is a ubiquitin-like protein which plays an important role in cell cycle control and embryogenesis. Covalent attachment to its substrates requires prior activation by the E1 complex UBE1C-APPBP1 and linkage to the E2 enzyme UBE2M. Attachment of NEDD8 to cullins activates their associated E3 ubiquitin ligase activity, and thus promotes polyubiquitination and proteasomal degradation of cyclins and other regulatory proteins.
Specifications
Specifications
| Antigen | NEDD8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Nedd8 |
| Gene Accession No. | P29595, Q15843, Q71UE8 |
| Gene Alias | nedd8; NEDD-8; NEDD8 ubiquitin like modifier; nedd8.L; nedd8.S; neddylin; Neural precursor cell expressed developmentally down-regulated protein 8; neural precursor cell expressed, developmentally down-regulated 8; neural precursor cell expressed, developmentally down-regulated 8 L homeolog; neural precursor cell expressed, developmentally down-regulated 8 S homeolog; neural precursor cell expressed, developmentally down-regulated gene 8; neural precursor cell-expressed developmentally down-regulated; Rub1; si:zc14a17.5; ubiquitin; ubiquitin-like protein Nedd8; XELAEV_18007009mg |
| Gene Symbols | Nedd8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
