Learn More
Nedocromil, MedChemExpress
Product Code. 16485688
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16485688
Brand: Medchem Express HY13448
MedChemExpress Nedocromil suppresses the action or formation of multiple mediators, including histamine, leukotriene C4 (LTC4), and prostaglandin D2 (PGD2).
Nedocromil suppresses the action or formation of multiple mediators, including histamine, leukotriene C4 (LTC4), and prostaglandin D2 (PGD2).
Specifications
Chemical Name or Material Nedocromil
CAS 69049-73-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H17NO7
Quantity 10 mg
Synonym FPL 59002
Solubility Information DMSO : 16.67 mg/mL (44.89 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=CC3=C(N(CC)C(C(O)=O)=CC3=O)C(CCC)=C2O1)=O)O
Molecular Weight (g/mol) 371.34
Formula Weight 371.34
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only