Learn More
NEDP1/SENP8 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98854MFV500
Product Code. 18563681
  • € 428.00 / 0.10mL
Description
NEDP1/SENP8 Polyclonal antibody specifically detects NEDP1/SENP8 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mFluor Violet 500 SE
Immunohistochemistry-Paraffin
Rabbit
123228