missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDP1/SENP8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98854-100ul
This item is not returnable.
View return policy
Description
NEDP1/SENP8 Polyclonal antibody specifically detects NEDP1/SENP8 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NEDP1/SENP8 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| DEN1deneddylase-1, deneddylase 1, Deneddylase-1, EC 3.4.22.-, HsT17512, NEDD8-specific protease 1, NEDP1NEDD8 specific-protease cysteine 2, Protease, cysteine 2, protease, cysteine, 2 (NEDD8 specific), PRSC2, Sentrin/SUMO-specific protease SENP8, sentrin-specific protease 8, SUMO sentrin specific protease family member 8, SUMO/sentrin specific peptidase family member 8, SUMO/sentrin specific protease family member 8 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NEDP1/SENP8. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 123228 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction