missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDP1/SENP8 Antibody (OTI3E4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72931FR
This item is not returnable.
View return policy
Description
NEDP1/SENP8 Monoclonal specifically detects NEDP1/SENP8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| NEDP1/SENP8 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| SENP8 | |
| Full length recombinant protein of human NEDP1/SENP8 (NP_660205) produced in E.coli. | |
| 0.1 mL | |
| 123228.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3E4 | |
| Western Blot, Immunohistochemistry | |
| DEN1deneddylase-1, deneddylase 1, Deneddylase-1, EC 3.4.22.-, HsT17512, NEDD8-specific protease 1, NEDP1NEDD8 specific-protease cysteine 2, Protease, cysteine 2, protease, cysteine, 2 (NEDD8 specific), PRSC2, Sentrin/SUMO-specific protease SENP8, sentrin-specific protease 8, SUMO sentrin specific protease family member 8, SUMO/sentrin specific peptidase family member 8, SUMO/sentrin specific protease family member 8 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction