Learn More
B Braun™ Needle-Pro™ Hypodermic Safety Needles
Product Code. 10555200
Change view
Click to view available options
Needle Gauge:
21 G
26 G
30 G
Unit Size:
Pack of 100
Pack of 800
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge unitSize
10555200 26 G Pack of 100
12622105 21 G Pack of 100
15929330 30 G Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10555200 Supplier B Braun™ Supplier No. 4658301
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Needle-Pro devices are specifically designed to minimize the risk of needlestick injuries.
  • Designed to allow standard puncture technique without requiring extra steps
  • Luer slip 26 – 30 G devices compatible with Luer slip syringes
  • Luer lock and Luer slip 18 – 25 G devices compatible with both Luer slip and Luer lock syringes
  • Stainless steel cannula, PP sheath and hub
  • Visual and audible confirmation as the needle locks into place with a simple one-handed technique that keeps the user’s hand behind the needle at all times
  • In accordance with ISO 7864; color-coded hub conforms to ISO 6009
  • Provided in packs of 100
Specifications
Certifications/Compliance ISO 7864; ISO 6009
Material Polypropylene (Barrel), Polypropylene (Hub), Stainless Steel (Tip)
Color Brown
Needle Gauge 26 G
Diameter (Metric) Outer Needle 0.45 mm
For Use With (Application) Reduction of needlestick injuries
Length (Metric) 12 mm
Product Type Hypodermic safety needle
Quantity 100/Pk.
Connection Type Leur Slip