missing translation for 'onlineSavingsMsg'
Learn More

Neferine, MedChemExpress

Product Code. 16438988
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438988 10 mg 10mg
16481408 5 mg 5mg
16474837 50 mg 50mg
16463866 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16438988 Supplier Medchem Express Supplier No. HYN0441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neferine is a major bisbenzylisoquinline alkaloid. Neferine strongly inhibits NF-κB activation.

Neferine is a major bisbenzylisoquinline alkaloid. Neferine strongly inhibits NF-κB activation.

Specifications

Chemical Name or Material Neferine
CAS 2292-16-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C38H44N2O6
Quantity 10 mg
Synonym (-)-Neferine
Solubility Information DMSO : 125 mg/mL (200.07 mM; Need ultrasonic)
SMILES OC1=CC=C(C[C@@H]2N(C)CCC3=C2C=C(OC)C(OC)=C3)C=C1OC4=CC5=C(C=C4OC)CCN(C)[C@@H]5CC6=CC=C(OC)C=C6
Molecular Weight (g/mol) 624.77
Formula Weight 624.77
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.