missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEI3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-441SS
This item is not returnable.
View return policy
Description
NEI3 Polyclonal specifically detects NEI3 in Human samples. It is validated for Western Blot.
Specifications
| NEI3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NEIL3 | |
| Residues 164-177 (LRAESEVKKQKGRMLG) of the human NEI3 protein. 85% sequence identity to mouse and rat. | |
| Affinity Purified | |
| DNA Repair | |
| 55247 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 | |
| DNA glycosylase FPG2, DNA glycosylase hFPG2, DNA glycosylase/AP lyase Neil3, EC 3.2.2.-, EC 4.2.99.18, endonuclease 8-like 3, Endonuclease VIII-like 3, FGP2, FLJ10858, FPG2, hFPG2, hNEI3, nei endonuclease VIII-like 3 (E. coli), NEI3, Nei-like protein 3 | |
| Rabbit | |
| 67.7 kDa | |
| 0.025 mL | |
| Primary | |
| This is specific for human NEI3. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction