missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neisseria gonorrhoeae MOMP Antibody (386/418), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64715AF594
This item is not returnable.
View return policy
Description
Neisseria gonorrhoeae MOMP Monoclonal antibody specifically detects Neisseria gonorrhoeae MOMP in Bacteria samples. It is validated for ELISA.
Specifications
| Neisseria gonorrhoeae MOMP | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Major outer membrane proteins (MOMP) from a mixture of detergent-treated Neisseria gonorrhoeae strains: SB587, SB2262, SB2265 & SB2275. | |
| 0.1 mL | |
| Reacts with a 30-31.5 kD MOMP protein of N. gonorrhoeae. | |
| Store at 4°C in the dark. |
| ELISA | |
| 386/418 | |
| ELISA | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Bacteria | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction