Learn More
Invitrogen™ Neisseria gonorrhoeae MOMP Monoclonal Antibody (386/418)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183358
Description
The immunogen used to produce MA1-83358 is major outer membrane proteins (MOMP) from a mixture of detergent-treated Neisseria gonorrhoeae strains: SB587, SB2262, SB2265, & SB2275. Mouse anti Neisseria gonorrhoeae MOMP 30 kDa antibody, clone 386/418 reacts with the 30-31.5 kDa major outer membrane protein (MOMP) of Neisseria gonorrhoeae a gram-negative diplococcus known to be the causative agent of sexually transmitted gonorrhoea.
The genus Neisseria contains a number of species which are normal flora and pathogens of humans and animals. Of these species, the species of human origin, and particularly the pathogenic species, N. gonorrhoeae and N. meningitidis, have been studied extensively in an effort to control the infections they cause. Gonorrhea, caused by N. gonorrhoeae, is one of the most frequently reported infectious diseases in the United States and worldwide. Of the Neisseria and related species, only N. gonorrhoeae is considered always to be pathogenic i.e., cause disease. N. gonorrhoeae is not considered to be normal flora under any circumstances.
Specifications
| Neisseria gonorrhoeae MOMP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA | |
| 386/418 | |
| Unconjugated | |
| Gonococci; N. gonorrhoeae | |
| Major outer membrane proteins (MOMP) from a mixture of detergent-treated Neisseria gonorrhoeae strains: SB587, SB2262, SB2265, & SB2275. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.