missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36534AF350
This item is not returnable.
View return policy
Description
NEK1 Polyclonal antibody specifically detects NEK1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| NEK1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686D06121, DKFZp686K12169, EC 2.7.11, EC 2.7.11.1, KIAA1901MGC138800, Never in mitosis A-related kinase 1, NIMA (never in mitosis gene a)-related kinase 1, NimA-related protein kinase 1, NY-REN-55, protein-serine/threonine kinase, Renal carcinoma antigen NY-REN-55, serine/threonine-protein kinase Nek1, SRPS2 | |
| The immunogen recognized by the antibody maps to a region between residue 1208 to 1258 of human Never in Mitosis A-Related Kinase 1 using the numbering given in entry NP_036356.1. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 4750 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction