missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93636-0.02ml
This item is not returnable.
View return policy
Description
NEK1 Polyclonal antibody specifically detects NEK1 in Human samples. It is validated for Western Blot
Specifications
| NEK1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686D06121, DKFZp686K12169, EC 2.7.11, EC 2.7.11.1, KIAA1901MGC138800, Never in mitosis A-related kinase 1, NIMA (never in mitosis gene a)-related kinase 1, NimA-related protein kinase 1, NY-REN-55, protein-serine/threonine kinase, Renal carcinoma antigen NY-REN-55, serine/threonine-protein kinase Nek1, SRPS2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human NEK1 (NP_036356.1). MEKYVRLQKIGEGSFGKAILVKSTEDGRQYVIKEINISRMSSKEREESRREVAVLANMKHPNIVQYRESFEENGSLYIVMDYCEGGDLFKRINAQKGVLF | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 4750 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction