missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 307.65
Specifications
| Antigen | NEK10 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611706
|
Novus Biologicals
NBP2-99009-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633666
|
Novus Biologicals
NBP2-99009-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NEK10 Polyclonal antibody specifically detects NEK10 in Human samples. It is validated for Western BlotSpecifications
| NEK10 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| 0.2 μm filtered solution in PBS | |
| 152110 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ32685, NIMA (never in mitosis gene a)- related kinase 10 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NEK10. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.