missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK11 Antibody (OTI4H5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72933B
This item is not returnable.
View return policy
Description
NEK11 Monoclonal specifically detects NEK11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| NEK11 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| NEK11 | |
| Full length recombinant protein of human NEK11 (NP_079076) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI4H5 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.11, EC 2.7.11.1, FLJ23495, Never in mitosis A-related kinase 11, NIMA (never in mitosis gene a)- related kinase 11, NimA-related protein kinase 11, serine/threonine-protein kinase Nek11 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 79858.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction