Learn More
NEK11 Mouse anti-Human, Clone: OTI4H5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501583
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the never in mitosis gene A family of kinases. The encoded protein localizes to the nucleoli, and may function with NEK2A in the S-phase checkpoint. The encoded protein appears to play roles in DNA replication and response to genotoxic stress. Alternatively spliced transcript variants have been described.Specifications
| NEK11 | |
| Monoclonal | |
| Unconjugated | |
| NEK11 | |
| NIMA related kinase 11 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79858 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4H5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8NG66 | |
| NEK11 | |
| Full length human recombit protein of human NEK11 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.