missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK3 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98357B
This item is not returnable.
View return policy
Description
NEK3 Polyclonal specifically detects NEK3 in Mouse samples. It is validated for ELISA.
Specifications
| NEK3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Biotin | |
| EC 2.7.11, EC 2.7.11.1, glycogen synthase A kinase, HSPK 36, HSPK36, hydroxyalkyl-protein kinase, MGC29949, Never in mitosis A-related kinase 3, NIMA (never in mitosis gene a)-related kinase 3, NimA-related protein kinase 3, phosphorylase B kinase kinase, serine/threonine-protein kinase Nek3 | |
| Produced in rabbits immunized with purified, recombinant NEK3 (ABK42288.1; Met1-Ala509). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 4752 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction