missing translation for 'onlineSavingsMsg'
Learn More

NEK4 Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Brand:  Abnova PAB30509.100uL

Product Code. 16177900

  • € 619.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Rabbit polyclonal antibody raised against partial recombinant human NEK4.

  • High specificity
  • High reactivity
Specifications

Specifications

NEK4
Polyclonal
Rabbit polyclonal antibody raised against partial recombinant human NEK4.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
MGC33171/NRK2/STK2/pp12301
Rabbit
Antigen affinity purification
RUO
6787
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20-1:50) The optimal working dilution should be determined by the end user.
P51957
NEK4
Recombinant protein corresponding to human NEK4.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)