missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118888
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NEK6.
NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
Specifications
| NEK6 | |
| Polyclonal | |
| Unconjugated | |
| NEK6 | |
| 1300007C09Rik; Nek6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA related kinase 6; NIMA-related expressed kinase 6; NIMA-related kinase 6; nimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; serine/threonine-protein kinase Nek6; SID; SID6-1512 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10783, 360161, 59126 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59895, Q9ES70, Q9HC98 | |
| NEK6 | |
| A synthesized peptide derived from human NEK6(Accession Q9HC98), corresponding to amino acid residues S111-E161. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction