missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK6 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 149.35 - € 422.30
Specifications
| Antigen | NEK6 |
|---|---|
| Concentration | 0.1 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16806733
|
Proteintech
10378-1-AP-20UL |
20 μL |
€ 149.35
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16896723
|
Proteintech
10378-1-AP-150UL |
150 μL |
€ 422.30
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Aspergillus nidulans ′never in mitosis A′ (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.Specifications
| NEK6 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse, Non-human Primate | |
| P59895, Q9ES70, Q9HC98 | |
| 10783, 360161, 59126 | |
| NEK6 Fusion Protein Ag0266 | |
| Primary | |
| -20°C | |
| NEK6 |
| 0.1 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| NEK6, NimA related protein kinase 6, Protein kinase SID6 1512, SID6 1512 | |
| Nek6 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title