Learn More
Invitrogen™ NEK6 Recombinant Rabbit Monoclonal Antibody (24GB755)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557506
 View more versions of this product
Product Code. 30276461
  • € 564.00 / 100µL
Description
NEK6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NEK6
Recombinant Monoclonal
Unconjugated
NEK6
1300007C09Rik; Nek6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA related kinase 6; NIMA-related expressed kinase 6; NIMA-related kinase 6; nimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; serine/threonine-protein kinase Nek6; SID; SID6-1512
Rabbit
Affinity chromatography
RUO
10783, 360161, 59126
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB755
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P59895, Q9ES70, Q9HC98
NEK6
A synthesized peptide derived from human NEK6 (aa 250-313).
100 μL
Primary
Human, Mouse, Rat