missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK7 Antibody (S04-7A1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19713-50ul
This item is not returnable.
View return policy
Description
NEK7 Monoclonal antibody specifically detects NEK7 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NEK7 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S04-7A1 | |
| Unconjugated | |
| EC 2.7.11.1, Never in mitosis A-related kinase 7, NIMA (never in mitosis gene a)-related kinase 7, NimA-related protein kinase 7, serine/threonine-protein kinase Nek7 | |
| A synthetic peptide of human NEK7 (Uniprot # Q8TDX7) | |
| 50 μg | |
| Protein Kinase | |
| 140609 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction