Learn More
Invitrogen™ NEK7 Recombinant Rabbit Monoclonal Antibody (JE51-70)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA534877
Product Code. 17185543
  • € 445.00 / 100µL
Description
Positive Control: NIH/3T3, Jurkat, rat brain tissue, human liver carcinoma tissue, human placenta tissue, mouse skeletal muscle tissue, A549.
NEK7 (NIMA Related Kinase 7) is a Protein Coding gene. Diseases associated with NEK7 include Muckle-Wells Syndrome and Cinca Syndrome. Among its related pathways are Cell Cycle, Mitotic and Transport of the SLBP independent Mature mRNA. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is NEK6.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NEK7
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q8TDX7
NEK7
Recombinant protein within internal sequence of Human NEK7aa 200-302.
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
JE51-70
Unconjugated
NEK7
2810460C19Rik; AU020186; NEK7; Never in mitosis A-related kinase 7; NIMA (never in mitosis gene a)-related expressed kinase 7; NIMA (never in mitosis gene a)-related kinase 7; NIMA related kinase 7; NIMA-related expressed kinase 7; NIMA-related kinase 7; nimA-related protein kinase 7; Serine/threonine-protein kinase Nek7
Rabbit
Protein A
RUO
140609
-20°C, Avoid Freeze/Thaw Cycles, store in dark