missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93834-0.1ml
This item is not returnable.
View return policy
Description
NEK8 Polyclonal antibody specifically detects NEK8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| NEK8 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:500 | |
| EC 2.7.11, EC 2.7.11.1, JCKNIMA-related kinase 12a, MGC138445, NEK12A, nephrocystin 9, Never in mitosis A-related kinase 8, NIMA (never in mitosis gene a)- related kinase 8, Nima-related protein kinase 12a, NimA-related protein kinase 8, NPHP9NIMA-family kinase NEK8, serine/threonine-protein kinase Nek8, serine/thrionine-protein kinase NEK8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 270-390 of human NEK8 (NP_835464.1). GSVRMRRAEKSVAPSNTGSRTTSVRCRGIPRGPVRPAIPPPLSSVYAWGGGLGTPLRLPMLNTEVVQVAAGRTQKAGVTRSGRLILWEAPPLGAGGGSLLPGAVEQPQPQFISRFLEGQSG | |
| 0.1 mL | |
| Protein Kinase | |
| 284086 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction