missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK9 Antibody (OTI1H1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72939AF750
This item is not returnable.
View return policy
Description
NEK9 Monoclonal specifically detects NEK9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| NEK9 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NEK9 | |
| Full length recombinant protein of human NEK9 (NP_149107) produced in HEK293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1H1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| DKFZp434D0935, KIAA1995, MGC138306, MGC16714, NEK8, Nek8NERCC1, Nercc1 kinase, NERCCEC 2.7.11.1, Never in mitosis A-related kinase 9, NIMA (never in mitosis gene a)- related kinase 9, NimA-related kinase 8, NIMA-related kinase Nek8, NimA-related protein kinase 9, serine/threonine-protein kinase Nek9 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, DNA Repair, GPCR, Phospho Specific | |
| 91754.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction