Learn More
Nelfinavir, MedChemExpress
Product Code. 16496418
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16496418
Brand: Medchem Express HY15287
MedChemExpress Nelfinavir (AG-1341) is a potent and orally bioavailable HIV-1 protease inhibitor (Ki=2 nM) for HIV infection. Nelfinavir is a broad-spectrum, anticancer agent.
Nelfinavir (AG-1341) is a potent and orally bioavailable HIV-1 protease inhibitor (Ki=2 nM) for HIV infection. Nelfinavir is a broad-spectrum, anticancer agent.
Specifications
Chemical Name or Material Nelfinavir
Physical Form Solid
CAS 159989-64-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C32H45N3O4S
For Use With (Application) COVID-19-anti-virus
Synonym AG1341
Solubility Information DMSO : ≥ 100 mg/mL (176.12 mM)
SMILES [H][C@]12C[C@@H](C(NC(C)(C)C)=O)N(C[C@H]([C@@H](NC(C3=C(C)C(O)=CC=C3)=O)CSC4=CC=CC=C4)O)C[C@@]1([H])CCCC2
Molecular Weight (g/mol) 567.78
Formula Weight 567.78
Percent Purity 96.9%
Grade Research
Show More Show Less
Research purposes only