Learn More
NELL2 Antibody (055), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90179JF549
Product Code. 18487918
  • € 607.00 / 0.10mL
Description
NELL2 Monoclonal antibody specifically detects NELL2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NELL2
055
This antibody was obtained from a rabbit immunized with purified, recombinant Human NELL2 (Uniprot#: Q99435; Met1-Leu816).
Monoclonal
Janelia Fluor 549