missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NELL2 Recombinant Rabbit Monoclonal Antibody (055)
GREENER_CHOICE

Product Code. 16344204
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16344204 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16344204 Supplier Invitrogen™ Supplier No. MA530700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human NELL2.

NELL2 is a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. Heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis.This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. An alternative splice variant has been described but its full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELL2
Applications ELISA
Classification Recombinant Monoclonal
Clone 055
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Nell2
Gene Accession No. Q99435
Gene Alias A330108N19Rik; LOW QUALITY PROTEIN: protein kinase C-binding protein NELL2; mel91; MEL91 protein; Nell2; NEL-like 2; NEL-like 2 (chicken); nel-like 2 homolog; NEL-like protein 2; Nel-related protein 2; neural EGFL like 2; neural epidermal growth factor-like 2; NRP2; Protein kinase C-binding protein NELL2; R75516
Gene Symbols Nell2
Host Species Rabbit
Immunogen Recombinant Human NELL2 protein (Met1-Leu816).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4753
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.