missing translation for 'onlineSavingsMsg'
Learn More

Neoandrographolide, MedChemExpress

Product Code. 16451488
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16451488

Brand: Medchem Express HYN0721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neoandrographolide is a diterpenoid from the Andrographis paniculata (Acanthaceae).

Neoandrographolide is a diterpenoid from the Andrographis paniculata (Acanthaceae).

Specifications

Chemical Name or Material Neoandrographolide
CAS 27215-14-1
Color White
Purity Grade Notes Research
Molecular Formula C26H40O8
Quantity 5 mg
Synonym Neoandrographiside
Solubility Information DMSO : 100 mg/mL (208.08 mM; Need ultrasonic)
SMILES C[C@@]([C@@H]1CCC2=CCOC2=O)(CCC3)[C@@](CCC1=C)([H])[C@]3(C)CO[C@@H]([C@@H]([C@@H](O)[C@@H]4O)O)O[C@@H]4CO
Molecular Weight (g/mol) 480.59
Formula Weight 480.59
Percent Purity 99.65%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.