missing translation for 'onlineSavingsMsg'
Learn More

Weigert™ Neodisher LaboClean FLA

Product Code. 10630123
This item is not returnable. View return policy
Product Code. 10630123 Supplier Weigert™ Supplier No. 411226

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Removes stubborn residues on oil base and other organic contaminants. Weigert Neodisher LaboClean FLA is a universally applicable, concentrated cleaning agent with pronounced cleaning power. It is an alkaline liquid free of surfactants, phosphorus compounds and oxidizing agents.

LaboClean FLA is used for machine cleaning/disinfecting of laboratory equipment. It is Ideal for use in medical, biological, microbiological and chemical laboratories and is suitable for use with glass, stainless steel, plastic and ceramic equipment, including pharmacy glass and animal cages. (It is not suitable for anodized aluminum and other light metal alloys.)

Specifications:

  • Density: 1.4g/cm3 at 20°C
  • Viscosity: <50 mPa•s
  • Titration factor: 0.37
  • Dose: 2–6ml/l (depending on the degree of contamination of the flushing material)

Specifications

Form Concentrated Liquid
pH 12.5 to 12.9 (2 to 6 mL/L aq. soln., 20°C)
Appearance Clear Yellowish Brown
Composition 5–15% polycarboxylate, 25–50%potassium hydroxide
Container Type Cannister
Content And Storage Store at 0°C to 30°C for up to 3 years.
For Use With (Application) Machine cleaning/disinfecting of laboratory equipment made of glass, stainless steel, plastic and ceramic.
Volume (Metric) 25 kg
compliance-icons compliance-icons
Product Identifier
  • neodisher LaboClean FLA
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.