missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neogenin Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32272AF594
This item is not returnable.
View return policy
Description
Neogenin Polyclonal antibody specifically detects Neogenin in Human, Mouse samples. It is validated for Western Blot
Specifications
| Neogenin | |
| Polyclonal | |
| Western Blot | |
| HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGNDKFZp547A066 | |
| The immunogen this antibody was made to, maps to a region between residue 1411 to 1461 of human Neogenin using the numbering given in entry NP_002490.2 (GeneID 4756). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4756 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction