missing translation for 'onlineSavingsMsg'
Learn More

Neohesperidin dihydrochalcone, MedChemExpress

Product Code. 16459006
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459006 100 mg 100mg
16432356 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16459006 Supplier Medchem Express Supplier No. HYN0154
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Neohesperidin dihydrochalcone is a synthetic glycoside chalcone, is added to various foods and beverages as a low caloric artificial sweetener.

Neohesperidin dihydrochalcone is a synthetic glycoside chalcone, is added to various foods and beverages as a low caloric artificial sweetener.

Specifications

Chemical Name or Material Neohesperidin dihydrochalcone
CAS 20702-77-6
Color White
Purity Grade Notes Research
Molecular Formula C28H36O15
Quantity 100 mg
Synonym Neohesperidin DC NHDC
Solubility Information DMSO : 100 mg/mL (163.24 mM; Need ultrasonic)
SMILES O=C(C1=C(O)C=C(O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O[C@@]3([H])[C@@H]([C@@H]([C@H]([C@H](C)O3)O)O)O)C=C1O)CCC4=CC=C(OC)C(O)=C4
Molecular Weight (g/mol) 612.58
Formula Weight 612.58
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.