Learn More
(+)-Neomenthyl Acetate, TRC
Product Code. 30135304
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135304 2.5 g 2.5g
30011636 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135304 Supplier Toronto Research Chemicals Supplier No. TRCN3900372.5G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2552-91-2
Molecular Formula C12 H22 O2
Molecular Weight (g/mol) 198.3
Synonym D-Neomenthyl acetate, (1S,2S,5R)-2-isopropyl-5-methylcyclohexyl acetate
IUPAC Name [(1S,2S,5R)-2-isopropyl-5-methyl-cyclohexyl] acetate
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@@H]1OC(=O)C
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12+/m1/s1
Formula Weight 198.162
Chemical Name or Material (1S)-(+)-Neomenthyl acetate