missing translation for 'onlineSavingsMsg'
Learn More

Neoseptin 3, MedChemExpress

Product Code. 16490439
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490439 10 mg 10mg
16414106 25 mg 25mg
16402648 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490439 Supplier Medchem Express Supplier No. HYU00435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neoseptin 3 is a Toll-like receptor 4/myeloid differentiation factor 2 (mTLR4/MD-2) agonist with an EC50 of 18.5 μM.

Neoseptin 3 is a Toll-like receptor 4/myeloid differentiation factor 2 (mTLR4/MD-2) agonist with an EC50 of 18.5 μM.

Specifications

Chemical Name or Material Neoseptin 3
CAS 1622863-21-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H34N2O4
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (210.71 mM)
SMILES CC(C)(C)OC([C@H](CCC1=CC=CC=C1)NC(C2=CC(CCC3=CC=C(O)C=C3)=C(N)C=C2)=O)=O
Molecular Weight (g/mol) 474.59
Formula Weight 474.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.