missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nephrin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.00 - € 494.00
Specifications
| Antigen | Nephrin |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18153568
|
Novus Biologicals
NBP2-37834 |
0.1 mg |
€ 494.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609325
|
Novus Biologicals
NBP2-37834-0.025mg |
0.025 mg |
€ 178.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nephrin Polyclonal specifically detects Nephrin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Nephrin | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 4868 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human Nephrin. The immunogen is located within amino acids 870 - 920 of Nephrin. Amino Acid Squence: VPLDLQDPRYTEHTYH | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, Renal glomerulus-specific cell adhesion receptor | |
| NPHS1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title