missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neprilysin/CD10 Antibody (MME/8376R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20771CL1
This item is not returnable.
View return policy
Description
Neprilysin/CD10 Monoclonal antibody specifically detects Neprilysin/CD10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Neprilysin/CD10 | |
| Monoclonal | |
| CoraFluor 1 | |
| Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase | |
| Recombinant human Neprilysin/CD10 protein fragment (aa 1-500) (exact sequence is proprietary) | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
| 4311 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MME/8376R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction